CrossLab Maintenance Kits for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
01018-68722 5065-4498 5065-4499 G1310-68731 G1367-68730 01100-68000 01100-68001 G1310-68730 G1312-68730 G1313-68709 G1313-68730 G1310-68741 G1310-68742 G1312-68741 G1313-68719 G7129-68740 G1367-68734 G1367-68741 G4220-68742 G4226-68735 G5611-68741 G5667-68735 G7104-68741 G7120-68741 G7129-68730 G7167-68710 G7167-68730 01100-68002 01100-68003 5067-4699 5067-6652 G1329-68737 G4204-68741 G4220-68741 G4226-67001 G4280-68730 G4280-68770 G5668-68730 G1361-68710 5067-4113 5068-0319 G1376-68710 G4204-68742 G4226-68740 G4280-68710 G4280-68750 G7131-68740 G7137-68740 G7157-68730 G7161-68740 G7167-68720
CA01018-68722EA 1169.83 CAD
CA01018-68722 CA5065-4498 CA5065-4499 CAG1310-68731 CAG1367-68730 CA01100-68000 CA01100-68001 CA76515-176 CA76515-190 CA76515-202 CA76515-206 CAG1310-68741 CAG1310-68742 CAG1312-68741 CAG1313-68719 CAG7129-68740 CA8005-0915 CAG1367-68734 CAG1367-68741 CAG4220-68742 CAG4226-68735 CAG5611-68741 CAG5667-68735 CAG7104-68741 CAG7120-68741 CAG7129-68730 CAG7167-68710 CAG7167-68730 CAAG0110068002 CAAG0110068003 CAAG5067-4699 CAAG5067-6652 CAAGG132968737 CAAGG420468741 CAAGG422068741 CAAGG422667001 CAAGG428068730 CAAGG428068770 CAAGG566868730 CAGG1361-68710 CAAG5067-4113 CAAG5068-0319 CAAGG137668710 CAAGG420468742 CAAGG422668740 CAAGG428068710 CAAGG428068750 CAAGG713168740 CAAGG713768740 CAAGG715768730 CAAGG716168740 CAAGG716768720
CrossLab Maintenance Kits for HPLC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Maintenance kits with all the necessary parts for keeping your HPLC instrument at peak performance.
  • Lack of preventive maintenance can result in noisy baselines, shorter column life, lower analytical sensitivity, and variable retention times
  • Routine maintenance should be done on a regular basis to keep your Agilent LC system performing at its optimum
  • Agilent preventive maintenance kits ensure HPLC systems are at their best
  • Maintenance procedures can be performed all at once or as needed
Agilent provides a full range of HPLC maintenance kits thatMore Product Information