J&W CP-Wax 52 CB Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
CP8738I CP7668I CP7763I CP8723I CP7638I CP7658I CP7713II5 CP7743I CP7773I CP7773II5 CP7791I CP7792I CP8074I CP8553I CP8713I CP8746I CP8763I CP8773I CP8843I
CACP8738IEA 1372.38 CAD
CACP8738I CAAGCP7668I CAAGCP7763I CAAGCP8723I CAAGCP7638I CAAGCP7658I CAAGCP7713II5 CAAGCP7743I CAAGCP7773I CAAGCP7773II5 CAAGCP7791I CAAGCP7792I CAAGCP8074I CAAGCP8553I CAAGCP8713I CAAGCP8746I CAAGCP8763I CAAGCP8773I CAAGCP8843I
J&W CP-Wax 52 CB Columns, Agilent Technologies
Chromatography Columns
Highly polar polyethylene glycol phase that provides separations for a broad range of applications.
  • Polyethylene glycol (PEG) phase
  • High polarity, suitable for a broad range of separations
  • Excellent column-to-column inertness reproducibility
  • Sharp and symmetrical peak shape performance for active polar compounds
  • Bonded, crosslinked, and solvent rinsable
  • Wider temperature range than non-bonded polyethylene glycols
  • High resolution of low boiling point analytes
  • Excellent temperature stability for a variety of EPA and ASTM methods
  • QC tested with highly
More Product Information