CrossLab Tubing, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
1300502506 5021-7107 5180-4196 7157-0210 CP4006 CP4008 CP4025 CP4260 5062-2419 5190-6983 AL3000 AL3002 AL3003 CP4003 CP4005 CP4013 CP4024 CP4026 CP4032 CP4049 CP4056 R000383005
CA1300502506EA 597.11 CAD
CA1300502506 CA5021-7107 CA76514-722 CAAG7157-0210 CAAGCP4006 CAAGCP4008 CAAGCP4025 CAAGCP4260 CAAG5062-2419 CAAG5190-6983 CAAGAL3000 CAAGAL3002 CAAGAL3003 CAAGCP4003 CAAGCP4005 CAAGCP4013 CAAGCP4024 CAAGCP4026 CAAGCP4032 CAAGCP4049 CAAGCP4056 CAAGR000383005
CrossLab Tubing, Agilent Technologies
Tubing Chromatography Tubing
Copper, stainless steel, and UltiMetal Plus tubing for GC applications.
  • Copper tubing can be used for most applications, except those requiring the highest purity gas delivery system
  • Stainless steel and UltiMetal Plus tubing are appropriate for more demanding applications
Agilent GC tubing includes high-quality copper, stainless steel, and UltiMetal Plus tubing, which are manufactured and cleaned for chromatographic use. Agilent tubing comes in different lengths and depending on your flow requirements, is available in ¹/₁₆, ¹/₈, or ¹/₄". UltiMetalMore Product Information