CrossLab Thermal Desorption Supplies for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
MKI-MTD-1169 MKI-MTD-1204 5182-0946 C-AT010C C-CF020 C-DF010 C-GAT010C C-GT010 C-HY010C C-PL010 C-TBE10 C-TBME10 C-TBP1C1C C-TBP1TC C-UN010C MKI-C-CPLOK MKI-U-T1KORI MKI-UTD-5064 MKI-UTD-5105 MKI-UTE-5099 MKI-U-T11GPC-2S MKI-U-T12ME-2S MKI-U-T13DHS-2S MKI-U-T15ATA-2S MKI-U-T17O3P-2S MKI-U-T18325-2S MKI-U-T3ATX-2S MKI-SERUTD-5065
CAAGMKIMTD1169PK 613.98 CAD
CAAGMKIMTD1169 CAAGMKIMTD1204 CAAG5182-0946 CAAGC-AT010C CAAGC-CF020 CAAGC-DF010 CAAGC-GAT010C CAAGC-GT010 CAAGC-HY010C CAAGC-PL010 CAAGC-TBE10 CAAGC-TBME10 CAAGC-TBP1C1C CAAGC-TBP1TC CAAGC-UN010C CAAGMKICCPLOK CAAGMKIUT1KORI CAAGMKIUTD5064 CAAGMKIUTD5105 CAAGMKIUTE5099 CAGMKIUT11GPC2 CAGMKIUT12ME2S CAGMKIUT13DHS2 CAGMKIUT15ATA2 CAGMKIUT17O3P2 CAGMKIUT183252 CAGMKIUT3ATX2S CAGSERUTD5065
CrossLab Thermal Desorption Supplies for GC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Agilent offers the supplies you need for your Markes Thermal Desorber.
  • Quantitative sample recollection of all the split flows enables repeat analysis of critical samples, easy method validation, and overcomes the one-shot limitation of conventional TD systems
  • Electronic tube tagging (TubeTAG): RFID tube tags are available for industry standard sorbent tubes
  • Diffusion locking (DiffLok) for enhanced sample integrity and robust (mechanically simple) automation
  • Patented inert valving for compatibility with every TD application on a single analytical platform -
More Product Information