Bond Elut SCX Plates, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
A4960725
CAAGA4960725EA 700.43 CAD
CAAGA4960725
Bond Elut SCX Plates, Agilent Technologies
Microplates Solid Phase Extraction Plates (SPE)
Bond Elut SCX is a strong cation exchanger with a very low pKa and aromatic functionality.
  • Useful for compounds with cationic and nonpolar characteristics
  • Superior cleanup from a single sorbent
  • Very low pKa creates strong analyte interaction
Bond Elut SCX is a strong cation exchanger with a very low pKa. Although the pKa is similar to Bond Elut PRS, the presence of the benzene ring in the functional group increases the potential for nonpolar interactions. This nonpolar characteristic becomes important in ion exchange from aqueous systems.