Sample Racks & Overlays, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
6610025400 6610026600 G3286-80106 6610025500 6610026000 6610026100 6610026400 G3286-80103 G8410-40000 G8410-40001 6610026500 810166900 810167000 G8410-80111
CA6610025400EA 74.5 CAD
CA6610025400 CA6610026600 CAG3286-80106 CAAG6610025500 CAAG6610026000 CAAG6610026100 CAAG6610026400 CAAGG328680103 CAAGG841040000 CAAGG841040001 CAAG6610026500 CAAG810166900 CAAG810167000 CAAGG841080111
Sample Racks & Overlays, Agilent Technologies
Racks Tube Racks
Sample racks & overlays for SPS 4, SPS 3 & ASX 500 autosamplers for AAS, ICP-OES, MP-AES & ICP-MS.
  • Perfect for use in any lab. - meets the demands of daily usage in your lab.
  • Made of polypropylene - steam autoclavable at 121 °C (250 °F)
  • Available in different configurations to accommodate different sized vessels giving you sample handling flexibility
  • Each rack features a solid end plate, which can be used for labeling or rack/tube identification
  • Interlocking feet allow stacking of empty racks for storage
  • Can be placed into a water bath if
More Product Information