CTC Smart Autosampler Syringes, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
8010-1303 8010-1304 8010-1306 8010-1316 8010-1319 8010-1321 8010-1325 8010-1326 8010-1336 8010-1338 8010-1339 8010-1340 8010-1343 8010-1346 8010-1352
CAAG8010-1303EA 331.24 CAD
CAAG8010-1303 CAAG8010-1304 CAAG8010-1306 CAAG8010-1316 CAAG8010-1319 CAAG8010-1321 CAAG8010-1324 CAAG8010-1325 CAAG8010-1326 CAAG8010-1336 CAAG8010-1338 CAAG8010-1339 CAAG8010-1340 CAAG8010-1343 CAAG8010-1346 CAAG8010-1352
CTC Smart Autosampler Syringes, Agilent Technologies
Syringes Syringes for Chromatography
Agilent now offers Smart Syringes compatible with your PAL3 Autosampler!
  • Smart technology enables critical parameter tracking for each unique syringe
  • Compatible with all PAL3 models
  • CTC syringes provide precise and effective sample delivery from your CTC autosampler to Agilent GC
  • Handles both liquid and headspace injections
Agilent now offers smart syringes compatible with your PAL3 Autosampler! Each smart syringe is equipped with its own read/write chip that allow you to track critical parameters, such as the number of injections a specific syringeMore Product Information