Long Life Septa for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
5183-4762 5183-4761 5183-4761-100 8010-0239 8010-0240 8010-0233 8010-0234 8010-0241 8010-0242 8010-0245 8010-0246 8010-0251 8010-0255 8010-0256
CA5183-4762PK 204.86 CAD
CA5183-4762 CA76514-914 CA51834761100 CAAG8010-0239 CAAG8010-0240 CAAG8010-0233 CAAG8010-0234 CAAG8010-0241 CAAG8010-0242 CAAG8010-0245 CAAG8010-0246 CAAG8010-0251 CAAG8010-0255 CAAG8010-0256
Long Life Septa for GC, Agilent Technologies
Septa
With these premium nonstick septa, you'll never have to scrape bits of septum out of your inlet.
  • The preferred septum for autosamplers
  • Pre-pierced for extended life and reduced coring
  • Ideal for overnight runs; up to 400 injections per septum
  • Plasma coating eliminates sticking
  • Maximum injection port temperature 350 °C
  • Soft, 45 Durometer, easy on autosampler needles
  • Blister packaging for cleanliness and convenience
Agilent now treats each premium 11 mm inlet septa to eliminate sticking in the GC inlet. With the premium non-stick septa, you'll never haveMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable