Advanced Green Septa for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
5183-4760 5183-4759 5183-4759-100 5190-3158 8010-0208 8010-0201 8010-0206 8010-0207 8010-0202 8010-0203 8010-0204 8010-0205 8010-0209 8010-0210 8010-0211 8010-0212 8010-0213 8010-0214 8010-0215 8010-0216
CA5183-4760PK 166.28 CAD
CA5183-4760 CA76514-910 CA51834759100 CA5190-3158 CA8010-0208 CAAG8010-0201 CAAG8010-0206 CAAG8010-0207 CAAG8010-0202 CAAG8010-0203 CAAG8010-0204 CAAG8010-0205 CAAG8010-0209 CAAG8010-0210 CAAG8010-0211 CAAG8010-0212 CAAG8010-0213 CAAG8010-0214 CAAG8010-0215 CAAG8010-0216
Advanced Green Septa for GC, Agilent Technologies
Septa
Advanced green nonstick 11 mm septa eliminates sticking leading to scraping bits of spetum out of the GC inlet.
  • Our green septa provide more injections per septum
  • Plasma coating eliminates sticking in the injection port
  • Maximum injection port temperature: 350 °C
  • Features blister packaging for cleanliness and convenience
Agilent now coats each premium 11 mm inlet septa with plasma to eliminate sticking in the GC inlet. With our premium non-stick septa, you'll never have to scrape bits of septum out of your inlet. Our premium septa also have a recess onMore Product Information