CrossLab Inlet Liners for Third Party GC Instruments, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
8004-0104 8004-0114 8004-0156 8004-0170 8004-0178 8001-0157 8001-0160 8002-0160 8004-0152 8004-0165 8004-0167 8001-0103 8001-0104 8001-0106 8001-0151 8001-0153 8001-0154 8001-0158 8001-0159 8001-0162 8001-0163 8002-0101 8002-0102 8002-0103 8002-0104 8002-0151 8002-0152 8002-0153 8002-0154 8003-0105 8003-0107 8003-0153 8003-0158 8003-0159 8003-0160 8003-0166 8004-0101 8004-0102 8004-0103 8004-0106 8004-0107 8004-0108 8004-0109 8004-0110 8004-0111 8004-0113 8004-0116 8004-0119 8004-0151 8004-0153 8004-0154 8004-0155 8004-0158 8004-0159 8004-0160 8004-0161 8004-0162 8004-0163 8004-0164 8004-0168 8004-0171 8004-0176
CA8004-0104PK 171.43 CAD
CA8004-0104 CA8004-0114 CA8004-0156 CA8004-0170 CA8004-0178 CAAG8001-0157 CAAG8001-0160 CAAG8002-0160 CAAG8004-0152 CAAG8004-0165 CAAG8004-0167 CAAG8001-0103 CAAG8001-0104 CAAG8001-0106 CAAG8001-0151 CAAG8001-0153 CAAG8001-0154 CAAG8001-0158 CAAG8001-0159 CAAG8001-0162 CAAG8001-0163 CAAG8002-0101 CAAG8002-0102 CAAG8002-0103 CAAG8002-0104 CAAG8002-0151 CAAG8002-0152 CAAG8002-0153 CAAG8002-0154 CAAG8003-0105 CAAG8003-0107 CAAG8003-0153 CAAG8003-0158 CAAG8003-0159 CAAG8003-0160 CAAG8003-0166 CAAG8004-0101 CAAG8004-0102 CAAG8004-0103 CAAG8004-0106 CAAG8004-0107 CAAG8004-0108 CAAG8004-0109 CAAG8004-0110 CAAG8004-0111 CAAG8004-0113 CAAG8004-0116 CAAG8004-0119 CAAG8004-0151 CAAG8004-0153 CAAG8004-0154 CAAG8004-0155 CAAG8004-0158 CAAG8004-0159 CAAG8004-0160 CAAG8004-0161 CAAG8004-0162 CAAG8004-0163 CAAG8004-0164 CAAG8004-0168 CAAG8004-0171 CAAG8004-0176
CrossLab Inlet Liners for Third Party GC Instruments, Agilent Technologies
Chromatography Supplies Chromatography Inlet Liners
Agilent provides a range of high-quality inlet liners for various third-party GC systems.
  • The perfect mix of liner configurations and chemistries to solve a wide variety of application challenges
  • Agilent Ultra Inert liners offer reproducibility, robustness, and reliability
  • Agilent Ultra Inert liners are delivered in our exclusive touchless packaging
  • Agilent original deactivation is recommended for everyday analysis
Agilent inlet liners for third-party GC systems provide the optimum environment for samples to be vaporized and mixed with the carrier gasMore Product Information