Nebulizers and Supplies for PerkinElmer AAS Instruments, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
8003-0406 8003-0409 8003-0823 8003-0827 8003-0830 8003-0831 8003-0833 8003-0836 8003-0837 8003-0860 8003-0864
CAAG8003-0406EA 175.5 CAD
CAAG8003-0406 CAAG8003-0409 CAAG8003-0823 CAAG8003-0827 CAAG8003-0830 CAAG8003-0831 CAAG8003-0833 CAAG8003-0836 CAAG8003-0837 CAAG8003-0860 CAAG8003-0864
Nebulizers and Supplies for PerkinElmer AAS Instruments, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Agilent nebulizers and supplies are equivalent to PerkinElmer AA nebulizer components.
  • Guaranteed to be compatible with PerkinElmer AA instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Qualified on a genuine PerkinElmer system
  • Full portfolio of AA, ICP-OES, and ICP-MS consumables, standards, and services to ensure your PerkinElmer system runs at peak performance
  • Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system
More Product Information