Torches and Supplies for PerkinElmer ICP-MS Instruments, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
8003-0552 8003-0554 8003-0555 8003-0556 8003-0591 8003-0594
CAAG8003-0552EA 743.65 CAD
CAAG8003-0552 CAAG8003-0554 CAAG8003-0555 CAAG8003-0556 CAAG8003-0591 CAAG8003-0594
Torches and Supplies for PerkinElmer ICP-MS Instruments, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Torches for the PerkinElmer ICP-MS systems.
  • Guaranteed to be compatible with PerkinElmer ICP-MS instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
  • Full portfolio of AA, ICP-OES, and ICP-MS consumables, standards and services to ensure your PerkinElmer system runs at peak performance
  • Worldwide technical support team to answer questions regarding part fit, function, or application-specific questions and concerns to help you get the
More Product Information