CrossLab Cylindrical Cells for UV-Vis and UV-Vis-NIR Systems, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
6618000600 5061-3392 6610002200 6610002300
CA6618000600EA 389.23 CAD
CA6618000600 CAAG5061-3392 CAAG5063-6566 CAAG6610002200 CAAG6610002300
CrossLab Cylindrical Cells for UV-Vis and UV-Vis-NIR Systems, Agilent Technologies
Cuvettes Spectrophotometer Cuvettes
Agilent cylindrical cuvettes with 10 to 100 mm path length for both 8453/8454 and Cary UV systems.
  • Long pathlength, 2-port cylindrical cells provide extra sensitivity for low concentration samples
  • Micro cylindrical cells are for use with concentrated samples and help overcome high solvent absorbance (e.g. water in the NIR)
  • Requires a cylindrical cell holder for mounting in the optical path
  • Compatible with thermostatted cylindrical cell holder for added flexibility
  • All cylindrical cells are optimized for use with Agilent's market-leading portfolio of UV-Vis
More Product Information