Bond Elut Lipid Extraction Plates, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
5610-2043
CAAG5610-2043PK 2597.91 CAD
CAAG5610-2043
Bond Elut Lipid Extraction Plates, Agilent Technologies
Microplates Solid Phase Extraction Plates (SPE)
Bond Elut Lipid Extraction SPE 1 mL cartridges and 2 mL 96-well plates provide highly selective and efficient extraction of lipids for lipid analysis, lipid profiling, and lipidomics workflows.
  • Streamlined sample preparation for lipid analysis
  • Features innovative EMR-Lipid sorbent technology
  • Time savings compared to traditional liquid-liquid extraction methods
  • mproved reproducibility compared to traditional liquid-liquid extraction methods, reducing the number of samples required for statistical evaluation
  • 2 ml well plate design is easily automatable for
More Product Information