Capillaries for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
01048-87302 01078-87305 01090-87304 01090-87308 01090-87610 5021-1831 5065-9927 5065-9930 5065-9932 5065-9933 5065-9935 5065-9979 5065-9980 79826-87606 79826-87607 79835-87638 79841-87609 79841-87610 79846-87605 G1312-67301 G1312-67500 G1312-87300 G1312-87305 G1312-87306 G1313-87304 G1313-87307 G1313-87308 G1315-87307 G1315-87312 G1316-27301 G1316-87304 G1316-87305 G1316-87306 G1316-87309 G1316-87312 G1316-87313 G1316-87314 G1316-87315 G1316-87316 G1316-87317 G1316-87318 G1316-87319 G1316-87323 G1316-87326 G1328-87600 G1361-67300 G1361-67301 G1361-67302 G1367-87302 G1375-87304 G1375-87306 G1375-87311 G1375-87312 G1375-87317 01090-87306 5021-1816 5021-1817 5021-1823 G1312-67305 G1315-87311 G1316-87300 01090-87611 5021-1818 5021-1819 5021-1820 5021-1821 5021-1822 5022-2159 5065-9931 G1312-67302 G1312-87301 G1312-87303 G1312-87304 G1313-87301 G1313-87305 G1316-87321 G1367-87303 5065-9963 5067-4655 5067-4656 5067-4659 5067-4669 5500-1157 5500-1217 5500-1243 5500-1246 G1329-87300 5065-9964 5067-4637 5067-4647 5067-4648 5067-4649 5067-4650 5067-4651 5067-4652 5067-4653 5067-4657 5067-4658 5067-4660 5067-4670 5067-4685 5067-4688 5067-4721 5067-4722 5067-4723 5067-4724 5067-4735 5067-4744 5067-4745 5067-4746 5067-4748 5067-5104 5067-5107 5067-5108 5067-5111 5067-5113 5067-5416 5067-6129 5500-1199 5500-1200 5500-1201 5500-1202 5500-1203 5500-1238 5500-1240 5500-1242 5500-1245 5500-1249 5500-1250 5500-1251 5500-1252 5500-1253 5500-1272 G1311-81600 G1311-81601 G1312-67300 G1315-87303 G1316-87328 G1367-87304 G4220-87000 G4226-60301 G5667-81001 G5667-81003 G5667-81004 G5667-81005 5022-6510 5023-2254 5067-1570 5067-1571 5067-4604 5067-4606 5067-4607 5067-4608 5067-4609 5067-4684 5067-4686 5067-4687 5067-4689 5067-4719 5067-4737 5067-4783 5067-5106 5067-5109 5067-5110 5067-5112 5067-5120 5067-5749 5067-5750 5067-5926 5067-6188 5067-6645 5067-6646 5067-6647 5067-6821 5500-1204 5500-1209 5500-1210 5500-1264 5500-1270 5500-1286 5500-1301 5500-1305 5500-1306 5500-1308 5500-1315 5500-1319 5500-1346 5500-1347 5500-1376 G1367-60301 G1375-87318 G1375-87320 G2258-87301 G2260-87300 G2260-87301 G5611-60500 G5611-60501 G5611-60502 G5611-60503 G5667-81000 G5667-81002 G5667-81007 5004-0016 5004-0017 5004-0018 5004-0019 5004-0041 5022-6508 5022-6509 5061-3361 5065-4493 5065-9906 5065-9926 5067-1572 5067-4605 5067-4784 5067-5709 5067-5713 5067-5939 5067-5941 5067-6643 5067-6644 5067-6820 5500-1278 5500-1280 5500-1284 5500-1300 5500-1302 5500-1303 5500-1309 5500-1314 5500-1316 5500-1317 5500-1318 5500-1320 5500-1329 5500-1330 5500-1333 5500-1334 5500-1335 5500-1336 5500-1384 5500-1385 5500-1386 5500-1387 5500-1388 5500-1390 5500-1391 5500-1393 5500-1398 5500-1406 5500-1407 5500-1408 5500-1415 5500-1423 5500-1432 5500-1433 5500-1434 5500-1438 5500-1441 5500-1442 5500-1444 5500-1445 5500-1447 5500-1451 5500-1456 5500-1462 79826-87601 79826-87605 79826-87608 G1313-87302 G1313-87306 G1315-68708 G1315-87305 G1315-87318 G1315-87323 G1315-87328 G1315-87333 G1315-87338 G1316-87327 G1361-67304 G1361-67305 G1361-67306 G1361-67500 G1361-67501 G1373-87300 G1375-87300 G1375-87301 G1375-87302 G1375-87303 G1375-87305 G1375-87308 G1375-87309 G1375-87310 G1375-87313 G1375-87314 G1375-87316 G1375-87319 G1375-87321 G1375-87322 G1375-87323 G1375-87324 G1375-87325 G1375-87327 G1375-87400 G1377-87301 G4240-87300 G4240-87301 G4240-87302 G4240-87303 G4240-87304 G4240-87310 G4280-81300 G4280-81301 G4301-60056 G5611-67300 G5611-67301 G5615-87302 G7132-60002
CA01048-87302EA 173.08 CAD
CA01048-87302 CA01078-87305 CA01090-87304 CA01090-87308 CA01090-87610 CA5021-1831 CA5065-9927 CA5065-9930 CA5065-9932 CA5065-9933 CA5065-9935 CA5065-9979 CA5065-9980 CA79826-87606 CA79826-87607 CA79835-87638 CA79841-87609 CA79841-87610 CA79846-87605 CAG1312-67301 CAG1312-67500 CAG1312-87300 CAG1312-87305 CAG1312-87306 CAG1313-87304 CAG1313-87307 CAG1313-87308 CAG1315-87307 CAG1315-87312 CAG1316-27301 CAG1316-87304 CAG1316-87305 CAG1316-87306 CAG1316-87309 CAG1316-87312 CAG1316-87313 CAG1316-87314 CAG1316-87315 CAG1316-87316 CAG1316-87317 CAG1316-87318 CAG1316-87319 CAG1316-87323 CAG1316-87326 CAG1328-87600 CAG1361-67300 CAG1361-67301 CAG1361-67302 CAG1367-87302 CAG1375-87304 CAG1375-87306 CAG1375-87311 CAG1375-87312 CAG1375-87317 CA76508-014 CA76508-034 CA76508-036 CA76508-038 CA76508-238 CA76508-246 CA76508-248 CA76514-514 CA76514-608 CA76514-610 CA76514-612 CA76514-614 CA76514-616 CA76514-620 CA76514-676 CA76515-188 CA76515-194 CA76515-196 CA76515-198 CA76515-208 CA76515-210 CA76515-214 CA76515-232 CA5065-9963 CA5067-4655 CA5067-4656 CA5067-4659 CA5067-4669 CA5500-1157 CA5500-1217 CA5500-1243 CA5500-1246 CAG1329-87300 CA5065-9964 CA5067-4637 CA5067-4647 CA5067-4648 CA5067-4649 CA5067-4650 CA5067-4651 CA5067-4652 CA5067-4653 CA5067-4657 CA5067-4658 CA5067-4660 CA5067-4670 CA5067-4685 CA5067-4688 CA5067-4721 CA5067-4722 CA5067-4723 CA5067-4724 CA5067-4735 CA5067-4744 CA5067-4745 CA5067-4746 CA5067-4748 CA5067-5104 CA5067-5107 CA5067-5108 CA5067-5111 CA5067-5113 CA5067-5416 CA5067-6129 CA5500-1199 CA5500-1200 CA5500-1201 CA5500-1202 CA5500-1203 CA5500-1238 CA5500-1240 CA5500-1242 CA5500-1245 CA5500-1249 CA5500-1250 CA5500-1251 CA5500-1252 CA5500-1253 CA5500-1272 CAG1311-81600 CAG1311-81601 CAG1312-67300 CAG1315-87303 CAG1316-87328 CAG1367-87304 CAG4220-87000 CAG4226-60301 CAG5667-81001 CAG5667-81003 CAG5667-81004 CAG5667-81005 CAAG5022-6510 CAAG5023-2254 CAAG5067-1570 CAAG5067-1571 CAAG5067-4604 CAAG5067-4606 CAAG5067-4607 CAAG5067-4608 CAAG5067-4609 CAAG5067-4684 CAAG5067-4686 CAAG5067-4687 CAAG5067-4689 CAAG5067-4719 CAAG5067-4737 CAAG5067-4783 CAAG5067-5106 CAAG5067-5109 CAAG5067-5110 CAAG5067-5112 CAAG5067-5120 CAAG5067-5749 CAAG5067-5750 CAAG5067-5926 CAAG5067-6188 CAAG5067-6645 CAAG5067-6646 CAAG5067-6647 CAAG5067-6821 CAAG5500-1204 CAAG5500-1209 CAAG5500-1210 CAAG5500-1264 CAAG5500-1270 CAAG5500-1286 CAAG5500-1301 CAAG5500-1305 CAAG5500-1306 CAAG5500-1308 CAAG5500-1315 CAAG5500-1319 CAAG5500-1346 CAAG5500-1347 CAAG5500-1376 CAAGG136760301 CAAGG137587318 CAAGG137587320 CAAGG225887301 CAAGG226087300 CAAGG226087301 CAAGG561160500 CAAGG561160501 CAAGG561160502 CAAGG561160503 CAAGG566781000 CAAGG566781002 CAAGG566781007 CAAG5004-0016 CAAG5004-0017 CAAG5004-0018 CAAG5004-0019 CAAG5004-0041 CAAG5022-6508 CAAG5022-6509 CAAG5061-3361 CAAG5065-4493 CAAG5065-9906 CAAG5065-9926 CAAG5067-1572 CAAG5067-4605 CAAG5067-4784 CAAG5067-5709 CAAG5067-5713 CAAG5067-5939 CAAG5067-5941 CAAG5067-6643 CAAG5067-6644 CAAG5067-6820 CAAG5500-1278 CAAG5500-1280 CAAG5500-1284 CAAG5500-1300 CAAG5500-1302 CAAG5500-1303 CAAG5500-1309 CAAG5500-1314 CAAG5500-1316 CAAG5500-1317 CAAG5500-1318 CAAG5500-1320 CAAG5500-1329 CAAG5500-1330 CAAG5500-1333 CAAG5500-1334 CAAG5500-1335 CAAG5500-1336 CAAG5500-1384 CAAG5500-1385 CAAG5500-1386 CAAG5500-1387 CAAG5500-1388 CAAG5500-1390 CAAG5500-1391 CAAG5500-1393 CAAG5500-1398 CAAG5500-1406 CAAG5500-1407 CAAG5500-1408 CAAG5500-1415 CAAG5500-1423 CAAG5500-1432 CAAG5500-1433 CAAG5500-1434 CAAG5500-1438 CAAG5500-1441 CAAG5500-1442 CAAG5500-1444 CAAG5500-1445 CAAG5500-1447 CAAG5500-1451 CAAG5500-1456 CAAG5500-1462 CAAG7982687601 CAAG7982687605 CAAG7982687608 CAAGG131387302 CAAGG131387306 CAAGG131568708 CAAGG131587305 CAAGG131587318 CAAGG131587323 CAAGG131587328 CAAGG131587333 CAAGG131587338 CAAGG131687327 CAAGG136167304 CAAGG136167305 CAAGG136167306 CAAGG136167500 CAAGG136167501 CAAGG137387300 CAAGG137587300 CAAGG137587301 CAAGG137587302 CAAGG137587303 CAAGG137587305 CAAGG137587308 CAAGG137587309 CAAGG137587310 CAAGG137587313 CAAGG137587314 CAAGG137587316 CAAGG137587319 CAAGG137587321 CAAGG137587322 CAAGG137587323 CAAGG137587324 CAAGG137587325 CAAGG137587327 CAAGG137587400 CAAGG137787301 CAAGG424087300 CAAGG424087301 CAAGG424087302 CAAGG424087303 CAAGG424087304 CAAGG424087310 CAAGG428081300 CAAGG428081301 CAAGG430160056 CAAGG561167300 CAAGG561167301 CAAGG561587302 CAAGG713260002
Capillaries for HPLC, Agilent Technologies
Tubing Chromatography Tubing
High-quality LC capillaries in a large variety of materials and dimensions.
  • A variety of top-quality materials available such as stainless steel, titanium, PEEK-sleeved stainless steel, and PEEK-coated fused silica
  • Agilent 0.6 mm od flexible 316 L grade stainless steel capillaries are corrosion resistant and much easier to handle than conventional rigid 1.6 mm od capillaries
  • Color-coded for easy identification of inner diameters
  • Bio-inert titanium or PEEK-sleeved stainless steel capillaries that fit perfectly for bio applications
All Agilent LCMore Product Information