Argon Humidifier Supplies for ICP-MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
5069-4496 5190-0148 G8400-60700
CAAG5069-4496EA 235.02 CAD
CAAG5069-4496 CAAG5190-0148 CAAGG840060700
Argon Humidifier Supplies for ICP-MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Conquer your challenging high TDS samples with the ICP-MS argon humidifier accessory and supplies.
  • Achieve accurate, reproducible analysis of samples containing elevated levels of total dissolved solids
  • Enables the accurate analysis of challenging sample types that may potentially cause particulate buildup, and eventually block the nebulizer, torch injector or interface cone orifice
  • Reduces frequency of routine maintenance when analyzing samples with elevated TDS
  • Significantly improve long-term stability for analysis of challenging samples
The AgilentMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable