CrossLab Flex Bench for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
5043-1735 8121-2258 5043-1245 5043-1278 5043-1289 5043-1725 5043-1759
CAAG5043-1735EA 441.14 CAD
CAAG5043-1735 CAAG8121-2258 CAAG5043-1245 CAAG5043-1278 CAAG5043-1289 CAAG5043-1725 CAAG5043-1759
CrossLab Flex Bench for HPLC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Easily configure your instrument to suit your needs anywhere in the lab.
  • Modify the height of your LC instrument, for easy access to solvent bottles and increased instrument optimization
  • Safely configure your instrument to suit the way you work
  • Move instruments where you need them, when you need them
  • Whether you want to work next to your MS or move analysts to new projects, the InfinityLab Flex Bench systems let you optimize your space while taking the hassle out of instrument relocation
  • Get more flexibility in how to set up your LC system
  • Special
More Product Information