CrossLab Fittings and Connectors for ICP-MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
5064-8017 5064-8019 G1820-65027 G3138-65129 5064-8022 5064-8023 5064-8024 5064-8026 5043-0054 G3280-67062 0100-2583 0100-2703 0535-1082 5023-1522 5023-1523 5023-1527 5042-4775 5043-0055 G1833-65430 G1833-65583 G3266-80010 G3266-80011 G3266-80013 G8400-60327 G8400-67026 5023-1517 5023-1519 5023-1520 5023-1521 5023-1524 5023-1525 5023-1526 5064-8018 5064-8021 5064-8027 5064-8078 G1820-60160 G1820-65052 G3138-65139 G3138-65140 G3138-65141 G3138-65143 G3138-65144 G3266-80017 G3666-20391
CA5064-8017EA 267.37 CAD
CA5064-8017 CA5064-8019 CAG1820-65027 CAG3138-65129 CA5064-8022 CA5064-8023 CA5064-8024 CA5064-8026 CA5043-0054 CAG3280-67062 CAAG0100-2583 CAAG0100-2703 CAAG0535-1082 CAAG5023-1522 CAAG5023-1523 CAAG5023-1527 CAAG5042-4775 CAAG5043-0055 CAAGG183365430 CAAGG183365583 CAAGG326680010 CAAGG326680011 CAAGG326680013 CAAGG840060327 CAAGG840067026 CAAG5023-1517 CAAG5023-1519 CAAG5023-1520 CAAG5023-1521 CAAG5023-1524 CAAG5023-1525 CAAG5023-1526 CAAG5064-8018 CAAG5064-8021 CAAG5064-8027 CAAG5064-8078 CAAGG182060160 CAAGG182065052 CAAGG313865139 CAAGG313865140 CAAGG313865141 CAAGG313865143 CAAGG313865144 CAAGG326680017 CAAGG366620391
CrossLab Fittings and Connectors for ICP-MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Fittings and connectors for Agilent 7700, 7800, 7900, 8800, 8900 ICP-MS including ISIS switching valve.
  • Find all fittings and connectors for your Agilent ICP-MS with easy filtering to identify the part you require.
  • Select by type, material and usage
  • Use genuine fittings and connectors on ICP-MS systems to ensure secure, leak free connections and trouble-free operation.
  • Preconfigured and pretested fittings and connectors simplify operation and maintenance for all Agilent ICP-MS instruments
Fittings and connectors for Agilent ICP-MS systems. From sampleMore Product Information