CrossLab Dissolution Paddles, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
14-3603 14-3595 14-3605 14-3608 13-3592 13-3593 13-3596 14-3590 14-3594 14-3597 14-3599 14-3600 14-3602 14-3604 14-3606 14-3607 16-3602 16-3603
CA14-3603EA 576.86 CAD
CA14-3603 CAAG14-3595 CAAG14-3605 CAAG14-3608 CAAG13-3592 CAAG13-3593 CAAG13-3596 CAAG14-3590 CAAG14-3594 CAAG14-3597 CAAG14-3599 CAAG14-3600 CAAG14-3602 CAAG14-3604 CAAG14-3606 CAAG14-3607 CAAG16-3602 CAAG16-3603
CrossLab Dissolution Paddles, Agilent Technologies
Dissolution Apparatus
Available in stainless steel, PTFE-coated, or PEEK in a range of sizes.
  • All paddles are serialized and designed to meet USP specifications for Apparatus 2
  • Select from electropolished stainless steel, PTFE-coated, or the biocompatible, chemically inert PEEK paddles in a range of sizes
  • Additionally, mini and mega paddles for small- and large-volume dissolution testing are available
All paddles are serialized and designed to meet USP specifications for USP Apparatus 2 including an individual CoC as standard. Paddles are available in stainless steel, PTFE-More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable