Bond Elut Cyano (CN), Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
12102034
CAAG12102034PK 314.94 CAD
CAAG12102034
Bond Elut Cyano (CN), Agilent Technologies
Chromatography Columns
Bond Elut Cyano is a medium-polarity phase available in endcapped and unendcapped forms.
  • Bond Elut (CN-E) ideal for extracting very nonpolar, aqueous analytes
  • Bond Elut (CN-U) retains polar compounds in nonpolar solvents
  • Suitable for many applications
Bond Elut Cyano (CN) is available as endcapped (CN-E) or unendcapped (CN-U) medium polarity sorbents. Bond Elut Cyano (CN-E) can be used effectively with extremely nonpolar analytes that would be irreversibly retained by C8 and C18 sorbents. This sorbent is best applied to aqueous samples.
Order Now
SPECIFICATIONS
Environmentally Preferable
Polarity Apolar stationary phase