2'-Deoxy-2'-fluorouridine 97%

Supplier: Thermo Scientific Chemicals
Warning

J65948-03 J65948-06 J65948-14
CAAAJ65948-03EA 55.3 CAD
CAAAJ65948-03 CAAAJ65948-06 CAAAJ65948-14
2'-Deoxy-2'-fluorouridine 97%
2'-Deoxy-2'-fluorouridine

Powder


Purity: 97% CAS No.: 784-71-4 Molecular Formula: C9H11FN2O5 Formula Weight: 246.19 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to off-white MDL No.: MFCD01317293

Formula: C₉H₁₁FN₂O₅
Storage Temperature: Ambient
MDL Number: MFCD01317293
CAS Number: 784-71-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR