DNA sodium salt (Deoxyribonucleic acid sodium salt) (from salmon testes)

Supplier: Thermo Scientific Chemicals

Synonyms: Thymonucleic acid sodium salt, DNA sodium salt

J65753-03 J65753-06
CAAAJ65753-03EA 243.32 CAD
CAAAJ65753-03 CAAAJ65753-06
DNA sodium salt (Deoxyribonucleic acid sodium salt) (from salmon testes)
DNA sodium salt (Deoxyribonucleic acid sodium salt)

solid


CAS No.: 68938-01-2 Storage Temperature: 2°C to 8°C Physical Form: solid Appearance: Off-white MDL No.: MFCD00284534

Storage Temperature: Ambient
MDL Number: MFCD00130921
CAS Number: 9007-49-2
Merck Index: 14,2907

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR