Tyrphostin C15

Supplier: Thermo Scientific Chemicals

J65227-MA
CAAAJ65227-MAEA 404.48 CAD
CAAAJ65227-MA
Tyrphostin C15
Tyrphostin C15

A selective ErbB2 inhibitor


Powder


Applications: A selective ErbB2 inhibitor

Formula: C₁₉H₁₅N₃O₃S₂
Storage Temperature: Refrigerator
MDL Number: MFCD01074971
CAS Number: 149092-50-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR