Isoluminol (4-aminophthalhydrazide) 98%

Supplier: Thermo Scientific Chemicals
Warning

J64921-03 J64921-06
CAAAJ64921-03EA 188.02 CAD
CAAAJ64921-03 CAAAJ64921-06
Isoluminol (4-aminophthalhydrazide) 98%
Isoluminol (4-Aminophthalhydrazide)

A chemiluminescent molecule.


Powder

Formula: C₈H₇N₃O₂
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00010560
CAS Number: 3682-14-2

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR