Tripeptidyl Peptidase Inhibitor

Supplier: Thermo Scientific Chemicals

J64760-LB0 J64760-MA
CAAAJ64760-LB0EA 197.5 CAD
CAAAJ64760-LB0 CAAAJ64760-MA
Tripeptidyl Peptidase Inhibitor
Proteins and Peptides

A serine protease inhibitor that acts as a potent, selective, dose-dependent, irreversible inhibitor of tripeptidylpeptidase II


CAS No.: 184901-82-4
Molecular Formula: C16H22ClN3O3
Formula Weight: 453.90
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Sensitivity: Light Sensitive
Appearance: Off-white
MDL No.: MFCD01310850

Order Now

SPECIFICATIONS

Protein/peptide name Tripeptidyl peptidase Inhibitor
Protein synonyms H-AAF-CMK
Environmentally Preferable
Species
Source
Storage conditions Freezer

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR