Caspase-3 Inhibitor III

Supplier: Thermo Scientific Chemicals

J64442-MCR
CAAAJ64442-LB0EA 1763.2 CAD
CAAAJ64442-LB0 CAAAJ64442-MCR
Caspase-3 Inhibitor III
Proteins and Peptides

A potent, cell-permeable, and irreversible inhibitor of caspase-3; also inhibits caspases -6, -7, -8, and -10. Commonly used at concentrations up to 100 M to examine the role of caspase-3-dependent apoptosis


  • Soluble in DMSO


CAS No.: 285570-60-7
Molecular Formula: C21H31ClN4O11
Formula Weight: 550.94
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: Off-white to brown
Solubility: Soluble in DMSO
MDL No.: MFCD01323001


Synonyms:
Ac-DEVD-CMK
N-Acetyl-Asp-Glu-Val-Asp chloromethyl ketone

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR