2'-Deoxyadenosine-5'-monophosphate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: dAMP, ((2R,3S,5R)-5-(6-Amino-9H-purin-9-yl)-3-hydroxytetrahydrofuran-2-yl)methyl dihydrogen phosphate

J64402-03 J64402-06
CAAAJ64402-03EA 152.31 CAD
CAAAJ64402-03 CAAAJ64402-06
2'-Deoxyadenosine-5'-monophosphate 98%
2'-Deoxyadenosine-5'-monophosphate

Crystalline powder


Purity: 98% CAS No.: 653-63-4 Molecular Formula: C10H14N5O6P Formula Weight: 331.22 Storage Temperature: 2°C to 8°C Physical Form: Crystalline powder Appearance: White MDL No.: MFCD00005753

Formula: C₁₀H₁₄N₅O₆P
MW: 331.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005753
CAS Number: 653-63-4
EINECS: 211-503-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR