Isradipine ≥98%

Supplier: Thermo Scientific Chemicals

J63920-MA J63920-MF
CAAAJ63920-MAEA 323.93 CAD
CAAAJ63920-MA CAAAJ63920-MF
Isradipine ≥98%
Isradipine

L-type calcium channel blocker


Crystalline powder


Applications: L-type calcium channel blocker

Formula: C₁₉H₂₁N₃O₅
Storage Temperature: Ambient
MDL Number: MFCD00153820
CAS Number: 75695-93-1
Merck Index: 14,5240

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR