3,5-Dimethyl-1-adamantanamine hydrochloride

Supplier: Thermo Scientific Chemicals
Warning

J63830-MF
CAAAJ63830-MFEA 257.18 CAD
CAAAJ63830-MF
3,5-Dimethyl-1-adamantanamine hydrochloride
3,5-Dimethyl-1-adamantanamine hydrochloride

An NMDA receptor antagonist; stimulates dopamine release


Powder


Applications: An NMDA receptor antagonist; stimulates dopamine release

Formula: C₁₂H₂₁N
Storage Temperature: Ambient
MDL Number: MFCD00214336
CAS Number: 41100-52-1
EINECS: 255-219-6
Merck Index: 14,5829

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR