Ketotifen fumarate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(1-Methylpiperidin-4-ylidene)-4H-benzo[4,5]cyclohepta[1,2-b]thiophen-10(9H)-one fumarate

J63708-03 J63708-MD
CAAAJ63708-03EA 410.94 CAD
CAAAJ63708-03 CAAAJ63708-MD
Ketotifen fumarate 99%
Ketotifen fumarate

A H1-histamine receptor antagonist that inhibits the anaphylactic release of histamine


Powder


Applications: A H1-histamine receptor antagonist that inhibits the anaphylactic release of histamine

Formula: C₁₉H₁₉NOS
MW: 425.50 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00079394
CAS Number: 34580-14-8
EINECS: 252-100-0
Merck Index: 14,5307

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR