Cetirizine dihydrochloride ≥99%

Supplier: Thermo Scientific Chemicals
Warning

J63549-06 J63549-09 J63549-14
CAAAJ63549-06EA 391.22 CAD
CAAAJ63549-06 CAAAJ63549-09 CAAAJ63549-14
Cetirizine dihydrochloride ≥99%
Cetirizine dihydrochloride

Histamine H-1 receptor antagonist. Inhibits activation of eosinophils and neutrophils


Solid


Applications: Histamine H-1 receptor antagonist. Inhibits activation of eosinophils and neutrophils

Formula: C₂₁H₂₅ClN₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00941428
CAS Number: 83881-52-1
Merck Index: 14,2022

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR