Calmidazolium chloride

Supplier: Thermo Scientific Chemicals

Synonyms: 1-(Bis(4-chlorophenyl)methyl)-3-(2-((2,4-dichlorobenzyl)oxy)-2-(2,4-dichlorophenyl)ethyl)-1H-imidazol-3-ium chloride

J63270-MA
CAAAJ63270-MAEA 743.37 CAD
CAAAJ63270-MA
Calmidazolium chloride
Calmidazolium chloride

Inhibits the calcium-calmodulin dependent phosphodiesterase


Solid


Applications: Inhibits the calcium-calmodulin dependent phosphodiesterase

Formula: C₃₁H₂₃Cl₇N₂O
MW: 687.71 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00077679
CAS Number: 57265-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR