Piroxicam

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Hydroxy-2-methyl-N-(2-pyridyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide, 4-Hydroxy-2-methyl-N-(pyridin-2-yl)-2H-benzo[e][1,2]thiazine-3-carboxamide 1,1-dioxide, 4-Hydroxy-2-methyl-3-(2-pyridylcarbamoyl)-2H-1,2-benzothiazine 1,1-Dioxide

J63239-03 J63239-06 J63239-09
CAAAJ63239-03EA 68.26 CAD
CAAAJ63239-03 CAAAJ63239-06 CAAAJ63239-09
Piroxicam
Piroxicam

A non-steroidal anti-inflammatory drug (NSAID) that is a highly selective inhibitor of cyclooxygenase-1 (COX-1)


Powder


Applications: A non-steroidal anti-inflammatory drug (NSAID) that is a highly selective inhibitor of cyclooxygenase-1 (COX-1)

Formula: C₁₅H₁₃N₃O₄S
MW: 331.35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00057317
CAS Number: 36322-90-4
EINECS: 252-974-3
UN: 2811
ADR: 6.1 III
Merck Index: 14,7506

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR