1-Butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide

Supplier: Thermo Scientific Chemicals
Danger

J62742-03 J62742-06 J62742-14
CAAAJ62742-03EA 284.4 CAD
CAAAJ62742-03 CAAAJ62742-06 CAAAJ62742-14
1-Butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide
1-Butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide

Sodium channel blocker


Cyrstalline powder


Applications: Sodium channel blocker

Formula: C₁₈H₂₈N₂O
Melting Pt: 106…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00243007
CAS Number: 38396-39-3
EINECS: 253-911-2
UN: 2811
ADR: "6.1",III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR