Stains-all 95%

Supplier: Thermo Scientific Chemicals
Warning

J62630-03 J62630-06
CAAAJ62630-03EA 143.34 CAD
CAAAJ62630-03 CAAAJ62630-06
Stains-all 95%
Stains-all

Cationic carbocyanine dye used in electrophoresis to stain proteins and nucleic acids


Powder


Applications: Cationic carbocyanine dye used in electrophoresis to stain proteins and nucleic acids

Formula: C₃₀H₂₇BrN₂S₂
Storage Temperature: Ambient
MDL Number: MFCD00078788
CAS Number: 7423-31-6
EINECS: 231-047-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR