Oxytetracycline hydrochloride

Supplier: Thermo Scientific Chemicals
Warning

J62489-22 J62489-14
CAAAJ62489-22EA 557.91 CAD
CAAAJ62489-22 CAAAJ62489-14
Oxytetracycline hydrochloride
Oxytetracycline hydrochloride

Impairs mitochondrial protein synthesis


Crystalline powder


Impairs mitochondrial protein synthesis

Formula: C₂₂H₂₅ClN₂O₉
Melting Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00135815
CAS Number: 2058-46-0
EINECS: 218-161-2
Merck Index: 14,6976

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR