Phosphate, 0.5 M Buffer Solution, pH 9.0, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J62345-AK J62345-AP
CAAAJ62345-AKEA 112.45 CAD
CAAAJ62345-AK CAAAJ62345-AP
Phosphate, 0.5 M Buffer Solution, pH 9.0, Thermo Scientific
Buffers Biological Buffers

Liquid


CAS No.: 7558-80-7
Physical Form: Liquid
MDL No.: MFCD00003527

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR