Adenosine-3',5'-cyclic monophosphate sodium salt 99%

Supplier: Thermo Scientific Chemicals

J62174-03 J62174-06 J62174-MD
CAAAJ62174-03EA 747.1 CAD
CAAAJ62174-03 CAAAJ62174-06 CAAAJ62174-MD
Adenosine-3',5'-cyclic monophosphate sodium salt 99%
Adenosine-3',5'-cyclic monophosphate sodium salt

Crystalline powder


Purity: 99% CAS No.: 37839-81-9 Molecular Formula: C10H11N5NaO6P Formula Weight: 351.19 Storage Temperature: -30°C to -10°C Physical Form: Crystalline powder Appearance: White Beilstein Reference: 54612 MDL No.: MFCD00069736

Formula: C₁₀H₁₁N₅NaO₆P
Storage Temperature: Freezer
MDL Number: MFCD00069736
CAS Number: 37839-81-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR