(-)-Epigallocatechin gallate

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (-)-cis-3,4-Dihydro-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-1(2H)-benzopyran-3-yl Gallate, (2R,3R)-5,7-Dihydroxy-2-(3,4,5-trihydroxyphenyl)chroman-3-yl 3,4,5-trihydroxybenzoate, EGCG

J61745-MF
CAAAJ61745-MFEA 277.05 CAD
CAAAJ61745-MF
(-)-Epigallocatechin gallate
(-)-Epigallocatechin gallate

An antioxidant that acts as an anti-promoter of carcinogenesis, inhibits angiogenesis and induces apoptosis


Powder


Applications: An antioxidant that acts as an anti-promoter of carcinogenesis, inhibits angiogenesis and induces apoptosis

Formula: C₂₂H₁₈O₁₁
MW: 458.38 g/mol
Melting Pt: 216…218 °C
Storage Temperature: Ambient
MDL Number: MFCD00075940
CAS Number: 989-51-5
Merck Index: 14,3526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR