L(+)-Histidine monohydrochloride monohydrate cell culture reagent

Supplier: Thermo Scientific Chemicals

J61465-09 J61465-22 J61465-36
CAAAJ61465-09EA 58.27 CAD
CAAAJ61465-09 CAAAJ61465-22 CAAAJ61465-36 CAAAJ61465-A1
L(+)-Histidine monohydrochloride monohydrate cell culture reagent
L(+)-Histidine monohydrochloride monohydrate

Powder


CAS No.: 5934-29-2 Molecular Formula: C6H9N3O2.HCl.H2O Formula Weight: 209.63 (191.62anhy) Physical Form: Powder Appearance: White Melting Point: ca 240° dec. Beilstein Reference: 4168261 Merck Reference: 14,4720 Optical Rotation: +10° (c=5 in 5N HCl) MDL No.: MFCD00151027

Formula: C₆H₉N₃O₂·HCl·H₂O
Melting Pt: ∼ 240 °C (dec.)
Density: 1.485 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00151027
CAS Number: 5934-29-2
EINECS: 211-438-9
Merck Index: 14,4720
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=561d2381f930afe618f78d680790ea9a43f6596c&vid=8aacf6dc4ef5382410cb743498272b48c3c36999&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR