Azure A

Supplier: Thermo Scientific Chemicals
Warning

J61346-14 J61346-22
CAAAJ61346-14EA 64.76 CAD
CAAAJ61346-14 CAAAJ61346-22
Azure A
Azure A

For staining cell granules and tissues.


Powder


For staining cell granules and tissues

Formula: C₁₄H₁₄ClN₃S
Melting Pt: 290 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012112
CAS Number: 531-53-3
EINECS: 208-510-7
Merck Index: 14,927

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR