Ciprofloxacin 98%

Supplier: Thermo Scientific Chemicals

J61317-06 J61317-14
CAAAJ61317-06EA 108.12 CAD
CAAAJ61317-06 CAAAJ61317-14
Ciprofloxacin 98%
Ciprofloxacin

An anti-bacterial agent effective against anaerobic bacteria


Powder


Applications: An anti-bacterial agent effective against anaerobic bacteria

Formula: C₁₇H₁₈FN₃O₃
Melting Pt: 253…257 °C
Storage Temperature: Freezer
MDL Number: MFCD00185755
CAS Number: 85721-33-1
Merck Index: 14,2314

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR