Calf Alkaline phosphatase (from Intestine)
Supplier: Thermo Scientific Chemicals
J61037-LB0 J61037-MCR
CAAAJ61037-LB0EA 2014.94 CAD
CAAAJ61037-LB0 CAAAJ61037-MCR
Calf Alkaline phosphatase (from Intestine)
Enzymes
MW = 140 kDa
Used to dephosphorylate proteins; also used to dephosphorylate the 5' termini of DNA or RNA to prevent self ligation.