SKF-96365 hydrochloride ≥99%

Supplier: Thermo Scientific Chemicals
Warning

J60937-MA
CAAAJ60937-MAEA 525.89 CAD
CAAAJ60937-MA CAAAJ60937-MF
SKF-96365 hydrochloride ≥99%
SKF-96365 hydrochloride

Inhibits receptor-mediated calcium influx


Powder


Applications: Inhibits receptor-mediated calcium influx

Formula: C₂₂H₂₆N₂O₃.HCl
Storage Temperature: Ambient
MDL Number: MFCD00236407
CAS Number: 130495-35-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR