Thymidine-5'-monophosphate disodium salt

Supplier: Thermo Scientific Chemicals

J60747-03 J60747-MC J60747-MD
CAAAJ60747-03EA 435.89 CAD
CAAAJ60747-03 CAAAJ60747-06 CAAAJ60747-MC CAAAJ60747-MD
Thymidine-5'-monophosphate disodium salt
Thymidine-5'-monophosphate disodium salt

Powder


CAS No.: 33430-62-5 Molecular Formula: C10H13N2Na2O8P Formula Weight: 366.18 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Solubility: Soluble in water Beilstein Reference: 4086927 MDL No.: MFCD09039259

Formula: C₁₀H₁₃N₂Na₂O₈P
Storage Temperature: Ambient
MDL Number: MFCD09039259
CAS Number: 33430-62-5
EINECS: 251-518-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR