Aminophylline, anhydrous 98%, powder

Supplier: Thermo Scientific Chemicals
Danger

J60705-14 J60705-22
CAAAJ60705-14EA 61.3 CAD
CAAAJ60705-14 CAAAJ60705-22
Aminophylline, anhydrous 98%, powder
Aminophylline

A non-selective phosphodiesterase inhibitor


Aminophylline exhibits vasodilatory and anti-inflammatory activity and is occasionally clinically used to treat chronic obstructive pulmonary disorder (COPD).


In animal models of allergen-induced lung inflammation, aminophylline decreases levels of IL-5, IL-8, and eosinophils. It is mostly used to prepare solutions for intravenous injections.

Formula: C₇H₈N₄O₂
Melting Pt: 269…274 °C
Storage Temperature: Freezer
MDL Number: MFCD00013221
CAS Number: 317-34-0
EINECS: 206-264-5
UN: 2811
ADR: "6.1",III
Merck Index: 14,465

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR