Hygromycin B

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (3aS,3'R,4S,4'S,5'R,6R,6'R,7S,7aS)-4-(((1R,2S,3R,5S,6R)-3-Amino-2,6-dihydroxy-5-(methylamino)cyclohexyl)oxy)-6'-((S)-1-amino-2-hydroxyethyl)-6-(hydroxymethyl)octahydro-3aH-spiro[[1,3]dioxolo[4,5-c]pyran-2,2'-pyran]-3',4',5',7-tetraol

J60681-03 J60681-MC J60681-MD
CAAAJ60681-03EA 297.04 CAD
CAAAJ60681-03 CAAAJ60681-MC CAAAJ60681-MD
Hygromycin B
Hygromycin B

An aminoglycoside antibiotic that kills bacteria by inhibiting protein synthesis


Lyophilized powder

Formula: C₂₀H₃₇N₃O₁₃
MW: 527.5 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06795479
CAS Number: 31282-04-9
EINECS: 250-545-5
UN: 3462
ADR: 6.1 III
Merck Index: 14,4852

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR