ACES, 0.5 M Buffer Solutions, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J60366-AE J60366-AK J60755-AE J60755-AK J61011-AK
CAAAJ60366-AEEA 170.01 CAD
CAAAJ60366-AE CAAAJ60366-AK CAAAJ60755-AE CAAAJ60755-AK CAAAJ61011-AE CAAAJ61011-AK CAAAJ61761-AE CAAAJ61761-AK
ACES, 0.5 M Buffer Solutions, Thermo Scientific
Buffers Biological Buffers

Liquid


CAS No.: 7365-82-4
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00008030

Order Now

SPECIFICATIONS

Environmentally Preferable
Physical form
CAS No.
MDL No. MFCD00008030
Appearance
Storage temperature

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR