1-Chloro-2-fluoro-3-nitrobenzene 97%

Supplier: Thermo Scientific Chemicals
Warning

H61989-14
CAAAH61989-14EA 108.23 CAD
CAAAH61989-14
1-Chloro-2-fluoro-3-nitrobenzene 97%
1-Chloro-2-fluoro-3-nitrobenzene

1-Chloro-2-fluoro-3-nitrobenzene, 97%

Formula: C₆H₃ClFNO₂
Storage Temperature: Ambient
MDL Number: MFCD00069417
CAS Number: 2106-49-2
EINECS: 218-285-7
UN: 2810
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR