Ethyl fluoroacetate 98%

Supplier: Thermo Scientific Chemicals
Danger

H61340-09 H61340-18
CAAAH61340-09EA 70.47 CAD
CAAAH61340-09 CAAAH61340-18
Ethyl fluoroacetate 98%
Ethyl fluoroacetate

Ethyl fluoroacetate, 98%

Formula: C₄H₇FO₂
Boiling Pt: 119…120 °C
Density: 1.093
Storage Temperature: Ambient
MDL Number: MFCD00000450
CAS Number: 459-72-3
EINECS: 207-297-8
UN: 2929
ADR: 6.1,II
Merck Index: 14,4167

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR